Anti-FBXW7 Mouse Monoclonal Antibody [clone: 1C11]

Supplier: Abnova
H00055294-M06
10585-830EA 502.5 CAD
10585-830
Anti-FBXW7 Mouse Monoclonal Antibody [clone: 1C11]
Antibodies
Type: Primary
Antigen: FBXW7 (F-box and WD repeat domain containing 7, E3 ubiquitin protein ligase)
Clonality: Monoclonal
Clone: 1C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR