Anti-GIMAP4 Mouse Monoclonal Antibody [clone: 1D8]

Supplier: Abnova
H00055303-M05
10587-362EA 502.5 CAD
10587-362
Anti-GIMAP4 Mouse Monoclonal Antibody [clone: 1D8]
Antibodies
Type: Primary
Antigen: GIMAP4 (GTPase, IMAP family member 4)
Clonality: Monoclonal
Clone: 1D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR