Anti-ZNF444 Mouse Monoclonal Antibody [clone: 4E9]

Supplier: Abnova
H00055311-M02
10605-916EA 502.5 CAD
10605-916
Anti-ZNF444 Mouse Monoclonal Antibody [clone: 4E9]
Antibodies
Type: Primary
Antigen: ZNF444
Clonality: Monoclonal
Clone: 4E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR