Anti-RNLS Mouse Polyclonal Antibody

Supplier: Abnova
H00055328-B01
10560-060EA 502.5 CAD
10560-060
Anti-RNLS Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RNLS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR