Anti-FBXL8 Rabbit Polyclonal Antibody

Supplier: Abnova
H00055336-D01P
10585-774EA 564 CAD
10585-774
Anti-FBXL8 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FBXL8 (F-box and leucine-rich repeat protein 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR