Anti-STK32B Mouse Monoclonal Antibody [clone: 4A7]

Supplier: Abnova
H00055351-M01
10602-738EA 502.5 CAD
10602-738
Anti-STK32B Mouse Monoclonal Antibody [clone: 4A7]
Antibodies
Type: Primary
Antigen: STK32B
Clonality: Monoclonal
Clone: 4A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR