Anti-STYK1 Mouse Monoclonal Antibody [clone: 3D2]

Supplier: Abnova
H00055359-M02
10602-908EA 502.5 CAD
10602-908
Anti-STYK1 Mouse Monoclonal Antibody [clone: 3D2]
Antibodies
Type: Primary
Antigen: STYK1
Clonality: Monoclonal
Clone: 3D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR