Anti-CHST12 Mouse Polyclonal Antibody

H00055501-B01P
10553-492EA 420.99 CAD
10553-492
Anti-CHST12 Mouse Polyclonal Antibody
Type: Primary
Antigen: CHST12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR