Anti-TNFRSF19 Mouse Monoclonal Antibody [clone: 1H6]

Supplier: Abnova
H00055504-M02
10604-054EA 502.5 CAD
10604-054
Anti-TNFRSF19 Mouse Monoclonal Antibody [clone: 1H6]
Antibodies
Type: Primary
Antigen: TNFRSF19
Clonality: Monoclonal
Clone: 1H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR