Anti-GPRC5D Mouse Polyclonal Antibody

Supplier: Abnova
H00055507-A01
10557-780EA 364.5 CAD
10557-780
Anti-GPRC5D Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GPRC5D (G protein-coupled receptor, class C, group 5, member D)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR