Anti-DDX43 Mouse Polyclonal Antibody

H00055510-B01P
10554-864EA 420.99 CAD
10554-864
Anti-DDX43 Mouse Polyclonal Antibody
Type: Primary
Antigen: DDX43
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR