Anti-FEM1A Mouse Polyclonal Antibody

Supplier: Abnova
H00055527-B01P
10556-518EA 502.5 CAD
10556-518
Anti-FEM1A Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FEM1A (fem-1 homolog a (C. elegans))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR