Anti-CDKN2AIP Mouse Monoclonal Antibody [clone: 4D11]

Supplier: Abnova
H00055602-M01
10578-640EA 502.5 CAD
10578-640
Anti-CDKN2AIP Mouse Monoclonal Antibody [clone: 4D11]
Antibodies
Type: Primary
Antigen: CDKN2AIP
Clonality: Monoclonal
Clone: 4D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR