Anti-PPP1R9A Mouse Monoclonal Antibody [clone: 6A10]

Supplier: Abnova
H00055607-M01
10597-904EA 502.5 CAD
10597-904
Anti-PPP1R9A Mouse Monoclonal Antibody [clone: 6A10]
Antibodies
Type: Primary
Antigen: PPP1R9A
Clonality: Monoclonal
Clone: 6A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR