Anti-VPS50 Mouse Monoclonal Antibody [clone: 2D11]

Supplier: Abnova
H00055610-M01
10586-218EA 502.5 CAD
10586-218
Anti-VPS50 Mouse Monoclonal Antibody [clone: 2D11]
Antibodies
Type: Primary
Antigen: VPS50
Clonality: Monoclonal
Clone: 2D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR