Anti-OTUB Mouse Monoclonal Antibody [clone: 1C12]

Supplier: Abnova
H00055611-M09
10595-840EA 502.5 CAD
10595-840
Anti-OTUB Mouse Monoclonal Antibody [clone: 1C12]
Antibodies
Type: Primary
Antigen: OTUB
Clonality: Monoclonal
Clone: 1C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR