Anti-TASP1 Mouse Monoclonal Antibody [clone: 3E10]

Supplier: Abnova
H00055617-M01
10578-294EA 502.5 CAD
10578-294
Anti-TASP1 Mouse Monoclonal Antibody [clone: 3E10]
Antibodies
Type: Primary
Antigen: TASP1
Clonality: Monoclonal
Clone: 3E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR