Anti-DEPDC1 Mouse Monoclonal Antibody [clone: 6H1]

Supplier: Abnova
H00055635-M05
10583-816EA 502.5 CAD
10583-816
Anti-DEPDC1 Mouse Monoclonal Antibody [clone: 6H1]
Antibodies
Type: Primary
Antigen: DEPDC1
Clonality: Monoclonal
Clone: 6H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR