Anti-CDC37L1 Mouse Monoclonal Antibody [clone: 1D4]

Supplier: Abnova
H00055664-M03
10581-638EA 502.5 CAD
10581-638
Anti-CDC37L1 Mouse Monoclonal Antibody [clone: 1D4]
Antibodies
Type: Primary
Antigen: CDC37L1
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR