Anti-ASF1B Mouse Monoclonal Antibody [clone: 4D5-2C2]

Supplier: Abnova
H00055723-M01
10577-372EA 502.5 CAD
10577-372
Anti-ASF1B Mouse Monoclonal Antibody [clone: 4D5-2C2]
Antibodies
Type: Primary
Antigen: ASF1B
Clonality: Monoclonal
Clone: 4D5-2C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR