Anti-CNDP2 Mouse Monoclonal Antibody [clone: 1A6]

Supplier: Abnova
H00055748-M10
10582-470EA 502.5 CAD
10582-470
Anti-CNDP2 Mouse Monoclonal Antibody [clone: 1A6]
Antibodies
Type: Primary
Antigen: CNDP2
Clonality: Monoclonal
Clone: 1A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR