Anti-TDP1 Mouse Monoclonal Antibody [clone: 2A10-G2]

Supplier: Abnova
H00055775-M01
10603-510EA 502.5 CAD
10603-510
Anti-TDP1 Mouse Monoclonal Antibody [clone: 2A10-G2]
Antibodies
Type: Primary
Antigen: TDP1
Clonality: Monoclonal
Clone: 2A10-G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR