Anti-MBD5 Mouse Monoclonal Antibody [clone: 4A12-1B6]

Supplier: Abnova
H00055777-M01
10592-990EA 502.5 CAD
10592-990
Anti-MBD5 Mouse Monoclonal Antibody [clone: 4A12-1B6]
Antibodies
Type: Primary
Antigen: MBD5
Clonality: Monoclonal
Clone: 4A12-1B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR