Anti-MBNL3 Mouse Polyclonal Antibody

H00055796-B01P
10563-876EA 420.99 CAD
10563-876
Anti-MBNL3 Mouse Polyclonal Antibody
Type: Primary
Antigen: MBNL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR