Anti-DCP1A Mouse Polyclonal Antibody

H00055802-B01P
10554-758EA 420.99 CAD
10554-758
Anti-DCP1A Mouse Polyclonal Antibody
Type: Primary
Antigen: DCP1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR