Anti-FOXJ2 Mouse Monoclonal Antibody [clone: 1G12]

Supplier: Abnova
H00055810-M09A
10623-968EA 502.5 CAD
10623-968
Anti-FOXJ2 Mouse Monoclonal Antibody [clone: 1G12]
Antibodies
Type: Primary
Antigen: FOXJ2 (forkhead box J2)
Clonality: Monoclonal
Clone: 1G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR