Anti-CENPN Mouse Monoclonal Antibody [clone: 4A5-1C11]

Supplier: Abnova
H00055839-M01
10581-172EA 502.5 CAD
10581-172
Anti-CENPN Mouse Monoclonal Antibody [clone: 4A5-1C11]
Antibodies
Type: Primary
Antigen: CENPN
Clonality: Monoclonal
Clone: 4A5-1C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR