Anti-LMO3 Mouse Monoclonal Antibody [clone: 4A8]

Supplier: Abnova
H00055885-M06A
10624-564EA 502.5 CAD
10624-564
Anti-LMO3 Mouse Monoclonal Antibody [clone: 4A8]
Antibodies
Type: Primary
Antigen: LMO3
Clonality: Monoclonal
Clone: 4A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR