Anti-MYNN Mouse Monoclonal Antibody [clone: 4B4]

Supplier: Abnova
H00055892-M02
10594-202EA 502.5 CAD
10594-202
Anti-MYNN Mouse Monoclonal Antibody [clone: 4B4]
Antibodies
Type: Primary
Antigen: MYNN
Clonality: Monoclonal
Clone: 4B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR