Anti-RNF114 Mouse Monoclonal Antibody [clone: 4G3-1A10]

Supplier: Abnova
H00055905-M01
10606-476EA 502.5 CAD
10606-476
Anti-RNF114 Mouse Monoclonal Antibody [clone: 4G3-1A10]
Antibodies
Type: Primary
Antigen: RNF114
Clonality: Monoclonal
Clone: 4G3-1A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR