Anti-CMAS Mouse Polyclonal Antibody

Supplier: Abnova
H00055907-A01
10553-694EA 364.5 CAD
10553-694
Anti-CMAS Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CMAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR