Anti-ERBB2IP Mouse Monoclonal Antibody [clone: 10G1]

Supplier: Abnova
H00055914-M01A
10624-272EA 502.5 CAD
10624-272
Anti-ERBB2IP Mouse Monoclonal Antibody [clone: 10G1]
Antibodies
Type: Primary
Antigen: ERBB2IP
Clonality: Monoclonal
Clone: 10G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR