Anti-ERBB2IP Mouse Monoclonal Antibody [clone: 10D2]

Supplier: Abnova
H00055914-M02
10584-624EA 502.5 CAD
10584-624
Anti-ERBB2IP Mouse Monoclonal Antibody [clone: 10D2]
Antibodies
Type: Primary
Antigen: ERBB2IP
Clonality: Monoclonal
Clone: 10D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR