Anti-APOM Mouse Monoclonal Antibody [clone: 4C7]

Supplier: Abnova
H00055937-M08
10577-160EA 502.5 CAD
10577-160
Anti-APOM Mouse Monoclonal Antibody [clone: 4C7]
Antibodies
Type: Primary
Antigen: APOM
Clonality: Monoclonal
Clone: 4C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR