Anti-BCAP29 Mouse Monoclonal Antibody [clone: 4B10]

Supplier: Abnova
H00055973-M07
10577-610EA 502.5 CAD
10577-610
Anti-BCAP29 Mouse Monoclonal Antibody [clone: 4B10]
Antibodies
Type: Primary
Antigen: BCAP29
Clonality: Monoclonal
Clone: 4B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR