Anti-PCDHGB2 Mouse Monoclonal Antibody [clone: 6E9]

Supplier: Abnova
H00056103-M02
10596-262EA 502.5 CAD
10596-262
Anti-PCDHGB2 Mouse Monoclonal Antibody [clone: 6E9]
Antibodies
Type: Primary
Antigen: PCDHGB2
Clonality: Monoclonal
Clone: 6E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR