Anti-PCDHB15 Mouse Monoclonal Antibody [clone: 2E10]

Supplier: Abnova
H00056121-M02
10596-246EA 502.5 CAD
10596-246
Anti-PCDHB15 Mouse Monoclonal Antibody [clone: 2E10]
Antibodies
Type: Primary
Antigen: PCDHB15
Clonality: Monoclonal
Clone: 2E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR