Anti-PCDHB14 Mouse Monoclonal Antibody [clone: 5F7]

Supplier: Abnova
H00056122-M01
10596-244EA 502.5 CAD
10596-244
Anti-PCDHB14 Mouse Monoclonal Antibody [clone: 5F7]
Antibodies
Type: Primary
Antigen: PCDHB14
Clonality: Monoclonal
Clone: 5F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR