Anti-PCDHAC2 Mouse Monoclonal Antibody [clone: 3D12]

Supplier: Abnova
H00056134-M03
10596-236EA 502.5 CAD
10596-236
Anti-PCDHAC2 Mouse Monoclonal Antibody [clone: 3D12]
Antibodies
Type: Primary
Antigen: PCDHAC2
Clonality: Monoclonal
Clone: 3D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR