Anti-TEX13A Rabbit Polyclonal Antibody

Supplier: Abnova
H00056157-D01P
10603-586EA 564 CAD
10603-586
Anti-TEX13A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TEX13A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR