Anti-CYP26B1 Mouse Monoclonal Antibody [clone: 2G7]

Supplier: Abnova
H00056603-M02
10583-372EA 502.5 CAD
10583-372
Anti-CYP26B1 Mouse Monoclonal Antibody [clone: 2G7]
Antibodies
Type: Primary
Antigen: CYP26B1
Clonality: Monoclonal
Clone: 2G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR