Anti-EIF5A2 Mouse Polyclonal Antibody

H00056648-B01P
10555-800EA 420.99 CAD
10555-800
Anti-EIF5A2 Mouse Polyclonal Antibody
Type: Primary
Antigen: EIF5A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR