Anti-TRIM39 Mouse Polyclonal Antibody

Supplier: Abnova
H00056658-B02
10572-954EA 502.5 CAD
10572-954
Anti-TRIM39 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TRIM39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR