Anti-JPH1 Mouse Monoclonal Antibody [clone: 2000000]

Supplier: Abnova
H00056704-M04
10590-892EA 502.5 CAD
10590-892
Anti-JPH1 Mouse Monoclonal Antibody [clone: 2000000]
Antibodies
Type: Primary
Antigen: JPH1
Clonality: Monoclonal
Clone: 2000000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR