Anti-RAD18 Mouse Monoclonal Antibody [clone: 2B9]

Supplier: Abnova
H00056852-M03
10599-044EA 502.5 CAD
10599-044
Anti-RAD18 Mouse Monoclonal Antibody [clone: 2B9]
Antibodies
Type: Primary
Antigen: RAD18
Clonality: Monoclonal
Clone: 2B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR