Anti-AGPAT4 Mouse Polyclonal Antibody

Supplier: Abnova

H00056895-B01
10510-508EA 502.5 CAD
10510-508
Anti-AGPAT4 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: AGPAT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR