Anti-EXOSC5 Mouse Monoclonal Antibody [clone: 1E11]

Supplier: Abnova
H00056915-M05
10585-414EA 502.5 CAD
10585-414
Anti-EXOSC5 Mouse Monoclonal Antibody [clone: 1E11]
Antibodies
Type: Primary
Antigen: EXOSC5
Clonality: Monoclonal
Clone: 1E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR