Anti-SEMA3G (sema Domain, immunoglobulin Domain (Ig), short basic Domain, secreted, (semaphorin) 3G) Mouse Polyclonal Antibody

H00056920-B01P
10570-836EA 420.99 CAD
10570-836
Anti-SEMA3G (sema Domain, immunoglobulin Domain (Ig), short basic Domain, secreted, (semaphorin) 3G) Mouse Polyclonal Antibody
Type: Primary
Antigen: SEMA3G (sema Domain, immunoglobulin Domain (Ig), short basic Domain, secreted, (semaphorin) 3G)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR