Anti-DUS3L Mouse Polyclonal Antibody

Supplier: Abnova
H00056931-B01
10555-444EA 502.5 CAD
10555-444
Anti-DUS3L Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DUS3L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR