Anti-CCNL1 Mouse Polyclonal Antibody

Supplier: Abnova
H00057018-B01
10552-758EA 502.5 CAD
10552-758
Anti-CCNL1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CCNL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR